Merck
  • Pharmacological inactivation of CHK1 and WEE1 induces mitotic catastrophe in nasopharyngeal carcinoma cells.
Pharmacological inactivation of CHK1 and WEE1 induces mitotic catastrophe in nasopharyngeal carcinoma cells.
Oncotarget (2015-05-31)
Joyce P Y Mak, Wing Yu Man, Jeremy P H Chow, Hoi Tang Ma, Randy Y C Poon
ABSTRACT
Nasopharyngeal carcinoma (NPC) is a rare but highly invasive cancer. As radiotherapy is the primary treatment for NPC, this offers a rationale to investigate if uncoupling the DNA damage responses can sensitize this cancer type. The G2 DNA damage checkpoint is controlled by a cascade of protein kinases: ATM/ATR, which phosphorylates CHK1/CHK2, which in turn phosphorylates WEE1. A number of small molecule inhibitors have been developed against these kinases as potential therapeutic agents. Here we demonstrated that compare to that in immortalized nasopharyngeal epithelial cells, ATR, CHK1, and WEE1 were overexpressed in NPC cell lines. Inhibitors of these kinases were unable to promote extensive mitotic catastrophe in ionizing radiation-treated NPC cells, indicating that they are not very effective radiosensitizer for this cancer. In the absence of prior irradiation, however, mitotic catastrophe could be induced with inhibitors against CHK1 (AZD7762) or WEE1 (MK-1775). NPC cells were more sensitive to WEE1 inactivation than nasopharyngeal epithelial cells. Targeting CHK1 and WEE1 together induced more extensive mitotic catastrophe than the individual components alone. Taken together, our results show that NPC cells depend on CHK1 and WEE1 activity for growth and that inhibitors of these kinases may serve as potential therapeutics for NPC.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
MISSION® esiRNA, targeting human CHEK1