Merck
  • Brain inflammation in a chronic epilepsy model: Evolving pattern of the translocator protein during epileptogenesis.
Brain inflammation in a chronic epilepsy model: Evolving pattern of the translocator protein during epileptogenesis.
Neurobiology of disease (2015-09-22)
Halima Amhaoul, Julie Hamaide, Daniele Bertoglio, Stephanie Nadine Reichel, Jeroen Verhaeghe, Elly Geerts, Debby Van Dam, Peter Paul De Deyn, Samir Kumar-Singh, Andrew Katsifis, Annemie Van Der Linden, Steven Staelens, Stefanie Dedeurwaerdere
ABSTRACT
A hallmark in the neuropathology of temporal lobe epilepsy is brain inflammation which has been suggested as both a biomarker and a new mechanistic target for treatments. The translocator protein (TSPO), due to its high upregulation under neuroinflammatory conditions and the availability of selective PET tracers, is a candidate target. An important step to exploit this target is a thorough characterisation of the spatiotemporal profile of TSPO during epileptogenesis. TSPO expression, microglial activation, astrocyte reactivity and cell loss in several brain regions were evaluated at five time points during epileptogenesis, including the chronic epilepsy phase in the kainic acid-induced status epilepticus (KASE) model (n = 52) and control Wistar Han rats (n = 33). Seizure burden was also determined in the chronic phase. Furthermore, ¹⁸F-PBR111 PET/MRI scans were acquired longitudinally in an additional four KASE animals. TSPO expression measured with in vitro and in vivo techniques was significantly increased at each time point and peaked two weeks post-SE in the limbic system. A prominent association between TSPO expression and activated microglia (p < 0.001; r = 0.7), as well as cell loss (p < 0.001; r = -0.8) could be demonstrated. There was a significant positive correlation between spontaneous seizures and TSPO upregulation in several brain regions with increased TSPO expression. TSPO expression was dynamically upregulated during epileptogenesis, persisted in the chronic phase and correlated with microglia activation rather than reactive astrocytes. TSPO expression was correlating with spontaneous seizures and its high expression during the latent phase might possibly suggest being an important switching point in disease ontogenesis which could be further investigated by PET imaging.
Product Description
Supelco
Diazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Diazepam
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%