Merck
  • Protein-Resistant Biodegradable Amphiphilic Graft Copolymer Vesicles as Protein Carriers.
Protein-Resistant Biodegradable Amphiphilic Graft Copolymer Vesicles as Protein Carriers.
Macromolecular bioscience (2015-06-04)
Yupeng Wang, Lesan Yan, Bin Li, Yanxin Qi, Zhigang Xie, Xiabin Jing, Xuesi Chen, Yubin Huang
ABSTRACT
The protein adsorption and self-assembly behavior of biocompatible graft copolymer, poly(lactide-co-diazidomethyl trimethylene carbonate)-g-poly(ethylene glycol) [P(LA-co-DAC)-g-PEG], were systematically studied. The graft copolymers showed enhanced resistance to non-specific protein adsorption compared with their block copolymer counterparts, indicative of the increased effect of PEG density beyond PEG length. Diverse nanostructures including vesicles can be assembled from the amphiphilic graft copolymers with well-defined nano-sizes. Hemoglobin (Hb), as a model protein, can be entrapped in the formed vesicles and keep the gas-binding capacity. The reduced release rate of Hb from graft copolymer vesicles indicated the relatively stable membrane packing compared with block copolymer counterpart.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%
Sigma-Aldrich
2-Ethylhexanoic acid, ≥99%
Sigma-Aldrich
2-Ethylhexanoic acid, 99%