Merck
  • 4,5-Substituted 3-Isoxazolols with Insecticidal Activity Act as Competitive Antagonists of Housefly GABA Receptors.
4,5-Substituted 3-Isoxazolols with Insecticidal Activity Act as Competitive Antagonists of Housefly GABA Receptors.
Journal of agricultural and food chemistry (2015-06-30)
Genyan Liu, Fumiyo Ozoe, Kenjiro Furuta, Yoshihisa Ozoe
ABSTRACT
The insect GABA receptor (GABAR), which is composed of five RDL subunits, represents an important target for insecticides. A series of 4,5-disubstituted 3-isoxazolols, including muscimol analogues, were synthesized and examined for their activities against four splice variants (ac, ad, bc, and bd) of housefly GABARs expressed in Xenopus oocytes. Muscimol was a more potent agonist than GABA in all four splice variants, whereas synthesized analogues did not exhibit agonism but rather antagonism in housefly GABARs. The introduction of bicyclic aromatic groups at the 4-position of muscimol and the simultaneous replacement of the aminomethyl group with a carbamoyl group at the 5-position to afford six 4-aryl-5-carbamoyl-3-isoxazolols resulted in compounds that exhibited significantly enhanced antagonism with IC50 values in the low micromolar range in the ac variant. The inhibition of GABA-induced currents by 100 μM analogues was approximately 1.5-4-fold greater in the ac and bc variants than in the ad and bd variants. 4-(3-Biphenylyl)-5-carbamoyl-3-isoxazolol displayed competitive antagonism, with IC50 values of 30, 34, 107, and 96 μM in the ac, bc, ad, and bd variants, respectively, and exhibited moderate insecticidal activity against houseflies, with an LD50 value of 5.6 nmol/fly. These findings suggest that these 3-isoxazolol analogues are novel lead compounds for the design and development of insecticides that target the orthosteric site of housefly GABARs.
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium carbonate, powder, 99.99% trace metals basis
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications