Merck
  • Cofilin 1 activation prevents the defects in axon elongation and guidance induced by extracellular alpha-synuclein.
Cofilin 1 activation prevents the defects in axon elongation and guidance induced by extracellular alpha-synuclein.
Scientific reports (2015-11-13)
Sharada Tilve, Francesco Difato, Evelina Chieregatti
ABSTRACT
Impaired adult neurogenesis and axon traumatic injury participate in the severity of neurodegenerative diseases. Alpha-synuclein, a cytosolic protein involved in Parkinson's disease, may be released from neurons, suggesting a role for excess secreted alpha-synuclein in the onset and spread of the pathology. Here we provide evidence that long term exposure of young neurons to extracellular alpha-synuclein hampers axon elongation and growth cone turning. We show that actin turnover and the rate of movement of actin waves along the axon are altered, due to alpha-synuclein-induced inactivation of cofilin. Upon laser disruption of microfilaments, healing of axons is favored by the increased phosphorylation of cofilin, however, at later time points; the defect in neurite extension prevails, being lost the regulation of cofilin activity. Importantly, overexpression of the active form of cofilin in neurons exposed to alpha-synuclein is able to restore the movement of actin waves, physiological axon elongation and growth cone turning. Our study reveals the molecular basis of alpha-synuclein-driven deficits in growth and migration of newborn neurons, and in elongation and regeneration of adult neurons.
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O