  • Search Results
  • Verification of chemical composition of commercially available propolis extracts by gas chromatography-mass spectrometry analysis.
Verification of chemical composition of commercially available propolis extracts by gas chromatography-mass spectrometry analysis.
Journal of medicinal food (2014-09-10)
Urszula Czyżewska, Joanna Konończuk, Joanna Teul, Paweł Drągowski, Renata Pawlak-Morka, Arkadiusz Surażyński, Wojciech Miltyk
ABSTRACT
Propolis is a resin that is collected by honeybees from various plant sources. Due to its pharmacological properties, it is used in commercial production of nutritional supplements in pharmaceutical industry. In this study, gas chromatography-mass spectrometry was applied for quality control analysis of the three commercial specimens containing aqueous-alcoholic extracts of bee propolis. More than 230 constituents were detected in analyzed products, including flavonoids, chalcones, cinnamic acids and their esters, phenylpropenoid glycerides, and phenylpropenoid sesquiterpenoids. An allergenic benzyl cinnamate ester was also identified in all tested samples. This analytical method allows to evaluate biological activity and potential allergenic components of bee glue simultaneously. Studies on chemical composition of propolis samples may provide new approach to quality and safety control analysis in production of propolis supplementary specimens.
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
N,O-Bis(trimethylsilyl)trifluoroacetamide, ≥99%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Hexane, anhydrous, 95%
Supelco
Methanol solution, suitable for HPLC, contains 10 % (v/v) water, 0.1 % (v/v) trifluoroacetic acid
Supelco
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Chlorotrimethylsilane, produced by Wacker Chemie AG, Burghausen, Germany, ≥99.0% (GC)
Sigma-Aldrich
Vinyl laurate, produced by Wacker Chemie AG, Burghausen, Germany, ≥98% (GC)
Sigma-Aldrich
Vinyl laurate, ≥99.0% (GC)
Sigma-Aldrich
Methanol, NMR reference standard