Merck
  • Chitin enhances serum IgE in Aspergillus fumigatus induced allergy in mice.
Chitin enhances serum IgE in Aspergillus fumigatus induced allergy in mice.
Immunobiology (2015-01-30)
Lalit Kumar Dubey, Jesper Bonnet Moeller, Anders Schlosser, Grith Lykke Sorensen, Uffe Holmskov
ABSTRACT
Aspergillus fumigatus (A. fumigatus) is a ubiquitous fungus that activates, suppresses or modulates the immune response by changing its cell wall structure and by secreting proteases. In this study, we show that chitin acts as an adjuvant in a murine model of A. fumigatus protease induced allergy. The mice were immunised intraperitoneally with A. fumigatus culture filtrate antigen either with or without chitin and were subsequently challenged with the culture filtrate antigen intranasally. Alum was used as an adjuvant control. Compared to alum, chitin induced a weaker inflammatory response in the lungs, measured as the total cell efflux in BAL, EPO and chitinase production. However, chitin enhanced the total IgE, specific IgE and specific IgG1 production as efficiently as alum. Pre-treatment with chitin but not with alum depressed the concentration of the Th2 cytokines IL-4 and IL-13 in BAL fluid. These results shows that chitin, in spite of a reduction of the Th2 cytokine levels in the lungs, enhanced the total and specific IgE production in A. fumigatus culture filtrate induced allergy.
Sigma-Aldrich
Chitinase Assay Kit, Fluorimetric, sufficient for 200 multiwell tests
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Water, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
E-Toxate Water, endotoxin, free
Sigma-Aldrich
Water, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Water, Molecular Biology, sterile filtered
Sigma-Aldrich
Water, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Water, PCR Reagent
Sigma-Aldrich
Water, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Water, BioPerformance Certified
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O
Sigma-Aldrich
2,3-Dimercapto-1-propanol, ≥98% (iodometric)