Merck
  • A Cholesterol-Dependent Endocytic Mechanism Generates Midbody Tubules During Cytokinesis.
A Cholesterol-Dependent Endocytic Mechanism Generates Midbody Tubules During Cytokinesis.
Traffic (Copenhagen, Denmark) (2015-09-25)
Emma Kettle, Scott L Page, Garry P Morgan, Chandra S Malladi, Chin L Wong, Ross A Boadle, Brad J Marsh, Phillip J Robinson, Megan Chircop
ABSTRACT
Cytokinesis is the final stage of cell division and produces two independent daughter cells. Vesicles derived from internal membrane stores, such as the Golgi, lysosomes, and early and recycling endosomes accumulate at the intracellular bridge (ICB) during cytokinesis. Here, we use electron tomography to show that many ICB vesicles are not independent but connected, forming a newly described ICB vesicular structure - narrow tubules that are often branched. These 'midbody tubules' labelled with horseradish peroxidase (HRP) within 10 min after addition to the surrounding medium demonstrating that they are derived from endocytosis. HRP-labelled vesicles and tubules were observed at the rim of the ICB after only 1 min, suggesting that midbody tubules are likely to be generated by local endocytosis occurring at the ICB rim. Indeed, at least one tubule was open to the extracellular space, indicative of a local origin within the ICB. Inhibition of cholesterol-dependent endocytosis by exposure to methyl-β-cyclodextrin and filipin reduced formation of HRP-labelled midbody tubules, and induced multinucleation following ICB formation. In contrast, dynamin inhibitors, which block clathrin-mediated endocytosis, induced multinucleation but had no effect on the formation of HRP-labelled midbody tubules. Therefore, our data reveal the existence of a cholesterol-dependent endocytic pathway occurring locally at the ICB, which contributes to the accumulation of vesicles and tubules that contribute to the completion of cytokinesis.
Sigma-Aldrich
Ethanol, purum, secunda spirit, denaturated with 2% 2-butanone, S15, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 2% 2-butanone, F25 MEK1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, suitable for hybridization, pH 7, ≥98%
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
RO-3306, ≥98% (HPLC)