Merck
  • 24 versus 48 Weeks of Peginterferon Plus Ribavirin in Hepatitis C Virus Genotype 6 Chronically Infected Patients with a Rapid Virological Response: A Non-Inferiority Randomized Controlled Trial.
24 versus 48 Weeks of Peginterferon Plus Ribavirin in Hepatitis C Virus Genotype 6 Chronically Infected Patients with a Rapid Virological Response: A Non-Inferiority Randomized Controlled Trial.
PloS one (2015-10-29)
Qingxian Cai, Xiaohong Zhang, Chaoshuang Lin, Xiaoqiong Shao, Yujuan Guan, Hong Deng, Min Wei, Mingshou Huang, Zefang Ren, Ling Lu, Yongyu Mei, Min Xu, Jianyun Zhu, Haiyan Shi, Guoli Lin, Ying Liu, Fengyu Hu, Qiumin Luo, Yun Lan, Fengxia Guo, Zhixin Zhao, Zhiliang Gao
ABSTRACT
The optimal treatment of hepatitis C virus (HCV) genotype 6 is unclear owing to its limited geographic distribution. Because of a high predictive value of rapid virological response (RVR) for sustained virological response (SVR), we conducted an open-label randomized controlled trial to compare 24- and 48-week peginterferon/ribavirin combination therapy for patients with HCV genotype 6 in Southern China who achieved an RVR. Treatment-naive, non-cirrhotic patients with chronic hepatitis C genotype 6 were treated with pegylated interferon α-2a (180 μg/week) and ribavirin (800-1,200 mg, according to weight) for 4 weeks. Patients who achieved an RVR, which was defined as HCV RNA negativity at week 4 (<50 IU), were randomized to receive either an additional 20 or 44 weeks of treatment (24- and 48-week treatment groups, respectively). The primary outcome measure was SVR. From January 2011 to June 2014, 152(152/210, 72.4%) patients with HCV genotype 6a and RVR were randomized 1:1 to the 24- or 48-week treatment group. The SVR rates in the 24- and 48-week groups in the intention-to-treat analysis were 90.8% (69/76) and 88.2% (67/76), respectively; those in the per-protocol analysis were 95.7% (67/70) and 97.0% (64/66), respectively. More patients in the 48-week group had anemia (46.1% vs. 28.9%, P = 0.03), but other adverse events were comparable between the groups. The limitation of the present study was that only patients from Southern China were enrolled which may inhibit the extensive application of the findings. Twenty-four weeks of peginterferon/ribavirin combination therapy was non-inferior to 48 weeks in patients with HCV genotype 6a in Southern China who achieved an RVR. ClinicalTrials.gov NCT01263860.
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)