Merck
  • Mechanism and Kinetics of Li2S Precipitation in Lithium-Sulfur Batteries.
Mechanism and Kinetics of Li2S Precipitation in Lithium-Sulfur Batteries.
Advanced materials (Deerfield Beach, Fla.) (2015-08-11)
Frank Y Fan, W Craig Carter, Yet-Ming Chiang
ABSTRACT
The kinetics of Li2 S electrodeposition onto carbon in lithium-sulfur batteries are characterized. Electrodeposition is found to be dominated by a 2D nucleation and growth process with rate constants that depend strongly on the electrolyte solvent. Nucleation is found to require a greater overpotential than growth, which results in a morphology that is dependent on the discharge rate.
Sigma-Aldrich
Sulfur, flakes, ≥99.99% trace metals basis
Sigma-Aldrich
Sulfur, powder, 99.98% trace metals basis
Sigma-Aldrich
Lithium-7Li, ≥99.8 atom % 7Li, ≥99.8% (CP)
Sigma-Aldrich
Lithium, granular, 4-10 mesh particle size, high sodium, 99% (metals basis)
Sigma-Aldrich
Diethylene glycol dimethyl ether, anhydrous, 99.5%
Sigma-Aldrich
Lithium sulfide, 99.98% trace metals basis
Sigma-Aldrich
sulfur powder, 99.998% trace metals basis
Sigma-Aldrich
Lithium, wire (in mineral oil), diam. 3.2 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, wire, diam. 3.2 mm, in mineral oil, ≥98%
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 19 mm, 99.9% trace metals basis
Sigma-Aldrich
Tetraethylene glycol dimethyl ether, ≥99%
Sigma-Aldrich
Lithium, ribbon, thickness × W 1.5 mm × 100 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, rod, diam. 12.7 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.75 mm × 45 mm, 99.9% trace metals basis
Sigma-Aldrich
Lithium, granular, 99% trace metals basis
Sigma-Aldrich
Lithium, ribbon, thickness × W 0.38 mm × 23 mm, 99.9% trace metals basis
Lithium, foil, not light tested, 38x200mm, thickness 0.20mm, as rolled, 99.9%
Sigma-Aldrich
Lithium, foil, not light tested, 38x500mm, thickness 0.20mm, as rolled, 99.9%
Lithium, foil, 25x100mm, thickness 0.6mm, as rolled, 99.9%