Merck
  • Analysis of porcine granulosa cell death signaling pathways induced by vinclozolin.
Analysis of porcine granulosa cell death signaling pathways induced by vinclozolin.
Theriogenology (2015-07-05)
Malgorzata Knet, Kamil Wartalski, Dorota Hoja-Lukowicz, Zbigniew Tabarowski, Maria Slomczynska, Malgorzata Duda
ABSTRACT
Recent studies suggest that disturbing androgen-signaling pathways in porcine ovarian follicles may cause granulosa cell (GC) death. For this reason, we investigated which apoptotic pathway is initiated after GC exposure to an environmental antiandrogen, vinclozolin (Vnz), in vitro. Immunocytochemistry, Western blots, and fluorometric assays were used to quantify caspase-3 and -9 expression and activity. To elucidate the specific mechanism of Vnz action and toxicity, GCs were assessed for viability, cytotoxicity, and apoptotic activity using the ApoTox-Glo Triplex Assay. To further determine the mechanism of GC death induced by Vnz, we used the Apoptosis Antibody Array Kit. In response to Vnz stimulus, we found an increased level of caspase-3 protein expression (P ≤ 0.001) and an increase in caspase-3 proteolytic activity (P ≤ 0.001), confirming that Vnz is a potent proapoptotic factor. The strong immunoreaction of caspase-9 after Vnz treatment (P ≤ 0.001) suggests that intrinsic mitochondrial apoptosis pathway was activated during GC death. On the other hand, caspase-8, being a part of the extrinsic receptor pathway, was also activated (P ≤ 0.001). Therefore, it is possible that Vnz induces porcine granulosal apoptosis also through a parallel pathway. Activation of these two pathways was confirmed by the Apoptosis Antibody Array Kit. In conclusion, it is possible that the intrinsic signaling pathway may not act as an initial trigger for GC apoptosis but might contribute to the amplification and propagation of apoptotic cell death in the granulosa layer after treatment with this antiandrogen. Moreover, Vnz disturbs the physiological process of programmed cell death. Consequently, this could explain why atretic follicles are rapidly removed and suggests that normal function of the ovarian follicle may be destroyed.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
3,3′-Diaminobenzidine, 97%
Sigma-Aldrich
3,3′-Diaminobenzidine, 97% (HPLC)
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
SAFC
Glycine
Sigma-Aldrich
N-Acetyl-Asp-Glu-Val-Asp-al, ≥95%, powder
Sigma-Aldrich
Trypan Blue, ≥80% (HPLC), Dye content 60 %
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Tris(tert-butoxy)silanol, 99.999%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%