Merck
  • Deproto-metallation of N-arylated pyrroles and indoles using a mixed lithium-zinc base and regioselectivity-computed CH acidity relationship.
Deproto-metallation of N-arylated pyrroles and indoles using a mixed lithium-zinc base and regioselectivity-computed CH acidity relationship.
Beilstein journal of organic chemistry (2015-10-02)
Mohamed Yacine Ameur Messaoud, Ghenia Bentabed-Ababsa, Madani Hedidi, Aïcha Derdour, Floris Chevallier, Yury S Halauko, Oleg A Ivashkevich, Vadim E Matulis, Laurent Picot, Valérie Thiéry, Thierry Roisnel, Vincent Dorcet, Florence Mongin
ABSTRACT
The synthesis of N-arylated pyrroles and indoles is documented, as well as their functionalization by deprotonative metallation using the base in situ prepared from LiTMP and ZnCl2·TMEDA (1/3 equiv). With N-phenylpyrrole and -indole, the reactions were carried out in hexane containing TMEDA which regioselectively afforded the 2-iodo derivatives after subsequent iodolysis. With pyrroles and indoles bearing N-substituents such as 2-thienyl, 3-pyridyl, 4-methoxyphenyl and 4-bromophenyl, the reactions all took place on the substituent, at the position either adjacent to the heteroatom (S, N) or ortho to the heteroatom-containing substituent (OMe, Br). The CH acidities of the substrates were determined in THF solution using the DFT B3LYP method in order to rationalize the experimental results.
Sigma-Aldrich
Pyrrole, reagent grade, 98%
Sigma-Aldrich
Indole, ≥99%
Sigma-Aldrich
Pyrrole, ≥98%, FCC, FG
Sigma-Aldrich
Indole, ≥99%, FG
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)