Merck
  • A new NMR approach for structure determination of thermally unstable biflavanones and application to phytochemicals from Garcinia buchananii.
A new NMR approach for structure determination of thermally unstable biflavanones and application to phytochemicals from Garcinia buchananii.
Magnetic resonance in chemistry : MRC (2015-07-22)
Timo D Stark, Sofie Lösch, Mathias Salger, Onesmo B Balemba, Junichiro Wakamatsu, Oliver Frank, Thomas Hofmann
ABSTRACT
Previous activity-guided phytochemical studies on Garcinia buchananii stem bark, which is traditionally used in Africa to treat various gastrointestinal and metabolic illnesses, revealed xanthones, polyisoprenylated benzophenones, flavanone-C-glycosides, biflavonoids, and/or biflavanones as bioactive key molecules. Unequivocal structure elucidation of biflavonoids and biflavanones by means of NMR spectroscopy is often complicated by the hindered rotation of the monomers around the C-C axis (atropisomerism), resulting in a high spectral complexity. In order to facilitate an unrestricted rotation, NMR spectra are usually recorded at elevated temperatures, commonly over 80 °C, which effects in a single set of resonance signals. However, under these conditions, one of the target compounds of this investigation, (2R,3S,2″R,3″R)-manniflavanone (1), undergoes degradation. Therefore, we demonstrated in the present study that the 1,1-ADEQUATE could be successfully used as a powerful alternative approach to confirm the C-C connectivities in 1, avoiding detrimental conditions. However, a moderate increase in temperature up to 50 °C was sufficient to deliver sharp signals in the proton NMR experiment of (2R,3S,2″R,3″R)-isomanniflavanone (2) and (2″R,3″R)-preussianone (3). In addition, two new compounds could be isolated, namely (2R,3S,2″R,3″R)-GB-2 7″-O-β-D-glucopyranoside (4) and (2R,3S,2″R,3″R)-manniflavanone-7″-O-β-D-glucopyranoside (5), and whose structures were elucidated by spectroscopic analysis including 1D and 2D NMR and mass spectrometry methods. The absolute configurations were determined by a combination of NMR and electronic circular dichroism (ECD) spectroscopy. The aforementioned compounds exhibited high anti-oxidative capacity in the H2O2 scavenging, hydrophilic Trolox equivalent antioxidant capacity (H-TEAC) and hydrophilic oxygen radical absorbance capacity (H-ORAC) assays.
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Deuterium oxide, filtered, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 70 atom % D
Sigma-Aldrich
Deuterium oxide, 60 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 1 % (w/w) 3-(trimethylsilyl)-1-propanesulfonic acid, sodium salt (DSS)
Sigma-Aldrich
Deuterium oxide, 99.8 atom % D
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
Sigma-Aldrich
Deuterium oxide, 99.9 atom % D