Merck
  • Ezrin interacts with the scaffold protein IQGAP1 and affects its cortical localization.
Ezrin interacts with the scaffold protein IQGAP1 and affects its cortical localization.
Biochimica et biophysica acta (2015-01-03)
Rathangadhara Chakrapani Nammalwar, Annika Heil, Volker Gerke
ABSTRACT
The cortical cytoskeleton constitutes an important subcellular structure that determines cell shape and regulates cell migration as well as membrane traffic to and from the plasma membrane. Many components of the cortical cytoskeleton have been identified including structural and scaffolding proteins, membrane-cytoskeleton linker proteins and signaling intermediates. We describe here an association of the membrane-F-actin linker protein ezrin with the scaffolding protein IQGAP1 that serves as a hub for concentrating different signaling complexes. Both, ezrin and IQGAP1 bind in a Ca²⁺-dependent manner to the EF hand protein S100P and complexes consisting of Ca²⁺-bound S100P, IQGAP1 and ezrin can be isolated by immunoprecipitation. Ezrin and IQGAP1 also interact in the absence of Ca²⁺, thus independent of S100P. Direct ezrin-IQGAP1 interaction can be shown with the purified proteins. It is mediated via the N-terminal FERM domain of ezrin and the IQ domain of IQGAP1, respectively. Ezrin and IQGAP1 colocalize in the submembraneous cytoskeleton and in cellular protrusions of human epithelial cells and knockdown of ezrin reduces the cortical localization of IQGAP1. Thus, ezrin appears to participate in recruiting IQGAP1 to the cell cortex thereby establishing a close connection between membrane-F-actin contacts and actin regulators that can be assembled by IQGAP1. This article is part of a Special Issue entitled: 13th European Symposium on Calcium.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Sigma-Aldrich
Calcium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
DL-Methionine, 99%, FCC, FG
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic acid, ~98%
Sigma-Aldrich
N-(2-Hydroxyethyl)ethylenediamine-N,N′,N′-triacetic acid, BioXtra, ≥98%
Sigma-Aldrich
DL-Methionine, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
DL-Methionine, ≥99%
Sigma-Aldrich
Calcium chloride
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ezr
Sigma-Aldrich
Calcium chloride, powder, 99.99% trace metals basis
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture