  • Search Results
  • Biodegradation of fat, oil and grease (FOG) deposits under various redox conditions relevant to sewer environment.
Biodegradation of fat, oil and grease (FOG) deposits under various redox conditions relevant to sewer environment.
Applied microbiology and biotechnology (2015-02-27)
Xia He, Qian Zhang, Michael J Cooney, Tao Yan
ABSTRACT
Fat, oil and, grease (FOG) deposits are one primary cause of sanitary sewer overflows (SSOs). While numerous studies have examined the formation of FOG deposits in sewer pipes, little is known about their biodegradation under sewer environments. In this study, FOG deposit biodegradation potential was determined by studying the biodegradation of calcium palmitate in laboratory under aerobic, nitrate-reducing, sulfate-reducing, and methanogenic conditions. Over 110 days of observation, calcium palmitate was biodegraded to CO2 under aerobic and nitrate-reducing conditions. An approximate 13 times higher CO2 production rate was observed under aerobic condition than under nitrate-reducing condition. Under sulfate-reducing condition, calcium palmitate was recalcitrant to biodegradation as evidenced by small reduction in sulfate. No evidence was found to support calcium palmitate degradation under methanogenic condition in the simulated sewer environment. Dominant microbial populations in the aerobic and nitrate-reducing microcosms were identified by Illumina seqeuncing, which may contain the capability to degrade calcium palmitate under both aerobic and nitrate-reducing conditions. Further study on these populations and their functional genes could shed more light on this microbial process and eventually help develop engineering solutions for SSOs control in the future.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Carbon, mesoporous, average pore diameter 100±10 Å (typical), >99.95% trace metals basis
Sigma-Aldrich
Carbon, mesoporous, nanopowder, <500 nm particle size (DLS), >99.95% trace metals basis
Sigma-Aldrich
Carbon, mesoporous, nanopowder, graphitized, <500 nm particle size (DLS), >99.95% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Carbon, mesoporous, hydrophilic pore surface
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Carbon, mesoporous
Sigma-Aldrich
Methane-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
Carbon nanofibers, graphitized (iron-free), composed of conical platelets, D × L 100 nm × 20-200 μm
Sigma-Aldrich
Carbon nanofibers, graphitized, platelets(conical), >98% carbon basis, D × L 100 nm × 20-200 μm
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foil, 100x100mm, thickness 2.0mm, glassy carbon
Carbon - Vitreous, foil, 8x8mm, thickness 0.5mm, glassy carbon