Merck
  • Antioxidant activity of protocatechuates evaluated by DPPH, ORAC, and CAT methods.
Antioxidant activity of protocatechuates evaluated by DPPH, ORAC, and CAT methods.
Food chemistry (2015-10-17)
Claudia Grajeda-Iglesias, Erika Salas, Nathalie Barouh, Bruno Baréa, Atikorn Panya, Maria Cruz Figueroa-Espinoza
ABSTRACT
Hibiscus sabdariffa L. is a worldwide consumed plant, principally after infusion of its dried sepals and calyces, which are usually discarded. Nevertheless, they represent a potential source of natural bioactive compounds, e.g. polyphenols, which could add value to this under-exploited plant. Protocatechuic acid (PA) was chosen as a model of the phenolic acids that can be extracted from H. sabdariffa. In order to modify PA hydrophilic character, which limits its use in lipid-rich food products, PA was esterified to C1-C18 alcohols, and the impact of lipophilization on its antioxidant activity was evaluated in both, an homogeneous (DPPH and ORAC methods) and an heterogeneous (CAT method) system. Results herein obtained showed that, depending on the grafted alkyl chain length, lipophilization could positively affect the antioxidant activity of PA in heterogeneous media; therefore, support its use as an innovative way to synthesize molecules with an improved antioxidant capacity and potential to be used as multifunctional preservatives in food.
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Lauryl alcohol, ≥98%, FG
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
1-Octadecanol, 95%
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
1-Hexanol, ReagentPlus®, ≥99.5% (GC)
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, purum, ≥99.0% (TLC)
Sigma-Aldrich
Fluorescein sodium salt, p.a.
Sigma-Aldrich
Fluorescein sodium salt, BioReagent, suitable for fluorescence
Sigma-Aldrich
3,4-Dihydroxybenzoic acid, ≥97.0% (T)
Sigma-Aldrich
Hexyl alcohol, FCC, FG
Sigma-Aldrich
1-Octadecanol, ReagentPlus®, 99%
Sigma-Aldrich
Fluorescein sodium salt, used as fluorescent tracer
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
5α-Androstan-17β-ol-3-one, ≥97.5%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanowires, diam. × L 2-6 nm × 200-400 nm
Sigma-Aldrich
Protocatechuic acid ethyl ester, 97%
Sigma-Aldrich
Hexyl alcohol, natural, ≥98%, FCC, FG
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)