Merck
  • Myocyte TLR4 enhances enteric and systemic inflammation driving late murine endotoxic ileus.
Myocyte TLR4 enhances enteric and systemic inflammation driving late murine endotoxic ileus.
American journal of physiology. Gastrointestinal and liver physiology (2015-03-10)
Bettina M Buchholz, Richard A Shapiro, Yoram Vodovotz, Timothy R Billiar, Chhinder P Sodhi, David J Hackam, Anthony J Bauer
ABSTRACT
Myocytes are nonhemopoietic in origin and functionally essential in generating gastrointestinal motility. In endotoxemia, a rapid-onset nonhemopoietic mechanism potently triggers early ileus in a Toll-like receptor 4 (TLR4)/myeloid differentiation primary response gene 88 (MyD88)-dependent manner. Moreover, synergistically with hemopoietic cells, nonhemopoietic cells escalate late ileus via an IL-6 receptor-dependent inflammation-driven pathway. We therefore specifically investigated the role of myocytes in TLR4-triggered inflammation and ileus. TLR4(+/+), TLR4(-/-), bmTLR4(+/+)/TLR4(-/-) chimera, SM22-Cre(-/-)TLR4(flox/flox), and selective myocyte TLR4-deficient (SM22-Cre(+/-)TLR4(flox/flox)) mice were injected intraperitoneally with purified lipopolysaccharide. SM22-driven Cre recombinase activity was selectively detected in cardiac, gastrointestinal, skeletal, and vascular myocytes, of small-sized vessels in a two-color fluorescent Cre reporter mouse. In contrast to nonhemopoietic TLR4 deficiency, deletion of myocyte TLR4 signaling prevented neither endotoxin-induced suppression of spontaneous jejunal contractility in vitro nor early ileus in vivo at 6 h. Circulating plasma colony-stimulating factor 3 was greatly elevated during endotoxemia, independent of myocyte TLR4 signaling or time. TLR4 activation of myocytes contributed significantly to an early enteric IL-6 mRNA induction and systemic IL-6 release, as well as to a late increase in circulating chemokine (C-X-C motif) ligand 1 (CXCL1) and IL-17. Consequently, inhibition of myocyte TLR4 signaling allowed functional recovery of motility by preventing inflammation-driven late ileus at 24 h. Direct TLR4 activation of myocytes is not responsible for nonhemopoietic-mediated early ileus. However, myocytes are proinflammatory cells that potently drive enteric and systemic inflammation, subsequently fueling late mediator-triggered ileus. Specifically, the myocyte TLR4-dependent inflammatory signature of elevated plasma IL-6, CXCL1, and IL-17 is strongly associated with late rodent ileus.
Sigma-Aldrich
Uracil, ≥99.0%
Sigma-Aldrich
Uracil, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)