Merck
  • Identification and Characterization of EDTA Test Strip Interfering Substances Using a Digital Color Detector.
Identification and Characterization of EDTA Test Strip Interfering Substances Using a Digital Color Detector.
Clinical laboratory (2015-08-25)
Kelly L Scholes, Ammon T Ence, Frederick G Strathmann, Jonathan R Genzen
ABSTRACT
Rapid test strips for ethylenediaminetetraacetic acid (EDTA) can be used to verify correct specimen types for clinical assays which require, or cannot be performed on, plasma collection tubes containing EDTA anticoagulant. As the test strip reaction chemistry is based on a color change induced by chelation of bismuth from a xylenol orange complex, we hypothesized that any agent capable of chelating bismuth might induce false positive test strip reactivity. The present study was therefore designed to evaluate the potential for test strip interference by chelating agents commonly used in the treatment of trace and heavy metal toxicity. A digital color detector mounted on a 3D-printed test strip holder was used to quantitatively assess test strip reactivity and evaluate concentration-response relationships of eight commercially available chelating agents. This approach revealed the following rank-order potency: K2EDTA = Na2EDTA > ethylene glycol tetra-acetic acid (EGTA) > dimercaptosuccinic acid (DMSA) > 2,3-dimercapto-1-propanesulfonic acid (DMPS) > penicillamine (PEN). Both deferoxamine (DEF) and alpha lipoic acid (ALA) were non-reactive at 10 mM concentrations. These experiments demonstrate that multiple substances can induce EDTA rapid test strip reactivity, but only at concentrations higher than might be expected during therapeutic chelation therapy. These agents are therefore unlikely to cause false positive results in routine clinical laboratory specimens.
Sigma-Aldrich
meso-2,3-Dimercaptosuccinic acid, ~98%
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, BioReagent, suitable for cell culture, powder, ≥98%
Sigma-Aldrich
5-Aminolevulinic acid hydrochloride, ≥98%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Citrate Concentrated Solution, BioReagent, suitable for coagulation assays, 4 % (w/v)
Sigma-Aldrich
Citrate Concentrated Solution, BioUltra, Molecular Biology, 1 M in H2O
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
(±)-α-Lipoic acid, synthetic, ≥99% (titration), powder
Sigma-Aldrich
(±)-α-Lipoic acid, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium 2,3-dimercaptopropanesulfonate monohydrate, 95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
(±)-α-Lipoic acid, ≥98.0%
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%