Merck
  • Versatile synthesis of 3,5-disubstituted 2-fluoropyridines and 2-pyridones.
Versatile synthesis of 3,5-disubstituted 2-fluoropyridines and 2-pyridones.
The Journal of organic chemistry (2003-04-12)
Andrew Sutherland, Timothy Gallagher
ABSTRACT
5-bromo-2-fluoro-3-pyridylboronic acid (3) was prepared in high yield by ortho-lithiation of 5-bromo-2-fluoropyridine (1), followed by reaction with trimethylborate. Suzuki reaction of 3 with a range of aryl iodides gave 3-monosubstituted 5-bromo-2-fluoropyridines 4 in excellent yields. A second Suzuki reaction utilizing the bromo constituent of 4 with aryl and heteroaryl boronic acids provided 3,5-disubstituted 2-fluoropyridines 5, which in turn could be converted to the corresponding 2-pyridones 6.
Sigma-Aldrich
5-Bromo-2-fluoropyridine, 99%
Pricing and availability is not currently available.