Merck
  • Copper(II) and nickel(II) binding sites of peptide containing adjacent histidyl residues.
Copper(II) and nickel(II) binding sites of peptide containing adjacent histidyl residues.
Journal of inorganic biochemistry (2015-07-21)
Ágnes Grenács, Daniele Sanna, Imre Sóvágó
ABSTRACT
Copper(II) and nickel(II) complexes of the terminally protected nonapeptide Ac-SGAEGHHQK-NH2 modeling the metal binding sites of the (8-16) domain of amyloid-β have been studied by potentiometric, UV-vis, CD and ESR spectroscopic methods. The studies on the mutants containing only one of the histidyl residues (Ac-SGAEGAHQK-NH2, Ac-SGAEGHAQK-NH2) have also been performed. The formation of imidazole and amide coordinated mononuclear complexes is characteristic of all systems with a preference of nickel(II) binding to the His14 site, while the involvement of both histidines in metal binding is suggested in the corresponding copper(II) complexes. The formation of bis(ligand) and dinuclear complexes has also been observed in the copper(II)-Ac-SGAEGHHQK-NH2 system. The results provide further support for the copper(II) binding ability of the (8-16) domain of amyloid-β and support the previous assumptions that via the bis(ligand) complex formation copper(II) ions may promote the formation of the oligomers of amyloid-β.
Sigma-Aldrich
2,2′-(Ethylenedioxy)diethanethiol, 95%