Merck
  • Progression of inflammation during immunodeficient mouse skeletal muscle regeneration.
Progression of inflammation during immunodeficient mouse skeletal muscle regeneration.
Journal of muscle research and cell motility (2015-11-29)
Iwona Grabowska, Magdalena A Mazur, K Kowalski, A Helinska, Jerzy Moraczewski, Władysława Stremińska, Grażyna Hoser, Jerzy Kawiak, Maria A Ciemerych, Edyta Brzoska
ABSTRACT
The skeletal muscle injury triggers the inflammatory response which is crucial for damaged muscle fiber degradation and satellite cell activation. Immunodeficient mice are often used as a model to study the myogenic potential of transplanted human stem cells. Therefore, it is crucial to elucidate whether such model truly reflects processes occurring under physiological conditions. To answer this question we compared skeletal muscle regeneration of BALB/c, i.e. animals producing all types of inflammatory cells, and SCID mice. Results of our study documented that initial stages of muscles regeneration in both strains of mice were comparable. However, lower number of mononucleated cells was noticed in regenerating SCID mouse muscles. Significant differences in the number of CD14-/CD45+ and CD14+/CD45+ cells between BALB/c and SCID muscles were also observed. In addition, we found important differences in M1 and M2 macrophage levels of BALB/c and SCID mouse muscles identified by CD68 and CD163 markers. Thus, our data show that differences in inflammatory response during muscle regeneration, were not translated into significant modifications in muscle regeneration.
Sigma-Aldrich
Pentobarbital sodium salt
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ptprc