Merck
  • piggyBac transposons expressing full-length human dystrophin enable genetic correction of dystrophic mesoangioblasts.
piggyBac transposons expressing full-length human dystrophin enable genetic correction of dystrophic mesoangioblasts.
Nucleic acids research (2015-12-20)
Mariana Loperfido, Susan Jarmin, Sumitava Dastidar, Mario Di Matteo, Ilaria Perini, Marc Moore, Nisha Nair, Ermira Samara-Kuko, Takis Athanasopoulos, Francesco Saverio Tedesco, George Dickson, Maurilio Sampaolesi, Thierry VandenDriessche, Marinee K Chuah
ABSTRACT
Duchenne muscular dystrophy (DMD) is a genetic neuromuscular disorder caused by the absence of dystrophin. We developed a novel gene therapy approach based on the use of the piggyBac (PB) transposon system to deliver the coding DNA sequence (CDS) of either full-length human dystrophin (DYS: 11.1 kb) or truncated microdystrophins (MD1: 3.6 kb; MD2: 4 kb). PB transposons encoding microdystrophins were transfected in C2C12 myoblasts, yielding 65±2% MD1 and 66±2% MD2 expression in differentiated multinucleated myotubes. A hyperactive PB (hyPB) transposase was then deployed to enable transposition of the large-size PB transposon (17 kb) encoding the full-length DYS and green fluorescence protein (GFP). Stable GFP expression attaining 78±3% could be achieved in the C2C12 myoblasts that had undergone transposition. Western blot analysis demonstrated expression of the full-length human DYS protein in myotubes. Subsequently, dystrophic mesoangioblasts from a Golden Retriever muscular dystrophy dog were transfected with the large-size PB transposon resulting in 50±5% GFP-expressing cells after stable transposition. This was consistent with correction of the differentiated dystrophic mesoangioblasts following expression of full-length human DYS. These results pave the way toward a novel non-viral gene therapy approach for DMD using PB transposons underscoring their potential to deliver large therapeutic genes.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid