Merck
  • Induction of the expressions of antioxidant enzymes by luteinizing hormone in the bovine corpus luteum.
Induction of the expressions of antioxidant enzymes by luteinizing hormone in the bovine corpus luteum.
The Journal of reproduction and development (2013-02-07)
Syota Kawaguchi, Ryosuke Sakumoto, Kiyoshi Okuda
ABSTRACT
Luteoprotective mechanisms of luteinizing hormone (LH) involved in the maintenance of bovine corpus luteum (CL) function have not been completely clarified. Since antioxidant enzymes are well documented as antiapoptotic factors in the CL of many mammals, we hypothesized that the luteoprotective action of LH is mediated by stimulating the local production and action of antioxidant enzymes. To test the above hypothesis, in the present study, we examined the mechanisms involved in the luteoprotective actions of LH. Cultured bovine luteal cells obtained from the CL at the mid-luteal stage (days 8-12 of the estrous cycle) were treated with LH (10 ng/ml), onapristone (OP; a specific progesterone receptor antagonist, 100 μM) and diethyldithiocarbamate [DETC; an inhibitor of superoxide dismutase (SOD), 100 μM] for 24 h. LH in combination with or without OP significantly increased the mRNA and protein expressions of manganese SOD (Mn-SOD) and catalase (CATA) and SOD activity. While LH alone significantly increased the mRNA and protein expressions of SOD containing copper and zinc (Cu,Zn-SOD), OP in combination with or without LH significantly decreased the mRNA and protein expressions of Cu,Zn-SOD. In addition, Cu,Zn-SOD, Mn-SOD and CATA mRNA expressions were higher at the mid luteal phase than the other luteal phases. LH in combination with DETC significantly decreased LH-increased cell viability. The overall results suggest that LH increases cell viability by LH-increased antioxidant enzymes, resulting in maintenance of CL function during the luteal phase in cattle.
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
N-Boc-1,3-propanediamine, ≥97.0% (GC/NT)
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, purified immunoglobulin, buffered aqueous solution