Merck
  • Identification of benzothiazoles as potential polyglutamine aggregation inhibitors of Huntington's disease by using an automated filter retardation assay.
Identification of benzothiazoles as potential polyglutamine aggregation inhibitors of Huntington's disease by using an automated filter retardation assay.
Proceedings of the National Academy of Sciences of the United States of America (2002-08-30)
Volker Heiser, Sabine Engemann, Wolfgang Bröcker, Ilona Dunkel, Annett Boeddrich, Stephanie Waelter, Eddi Nordhoff, Rudi Lurz, Nancy Schugardt, Susanne Rautenberg, Christian Herhaus, Gerhard Barnickel, Henning Böttcher, Hans Lehrach, Erich E Wanker
ABSTRACT
Preventing the formation of insoluble polyglutamine containing protein aggregates in neurons may represent an attractive therapeutic strategy to ameliorate Huntington's disease (HD). Therefore, the ability to screen for small molecules that suppress the self-assembly of huntingtin would have potential clinical and significant research applications. We have developed an automated filter retardation assay for the rapid identification of chemical compounds that prevent HD exon 1 protein aggregation in vitro. Using this method, a total of 25 benzothiazole derivatives that inhibit huntingtin fibrillogenesis in a dose-dependent manner were discovered from a library of approximately 184,000 small molecules. The results obtained by the filter assay were confirmed by immunoblotting, electron microscopy, and mass spectrometry. Furthermore, cell culture studies revealed that 2-amino-4,7-dimethyl-benzothiazol-6-ol, a chemical compound similar to riluzole, significantly inhibits HD exon 1 aggregation in vivo. These findings may provide the basis for a new therapeutic approach to prevent the accumulation of insoluble protein aggregates in Huntington's disease and related glutamine repeat disorders.
Sigma-Aldrich
PGL-135 hydrochloride monohydrate, ≥98% (HPLC), solid