Merck
  • Kindlin-1 controls Wnt and TGF-β availability to regulate cutaneous stem cell proliferation.
Kindlin-1 controls Wnt and TGF-β availability to regulate cutaneous stem cell proliferation.
Nature medicine (2014-04-01)
Emanuel Rognoni, Moritz Widmaier, Madis Jakobson, Raphael Ruppert, Siegfried Ussar, Despoina Katsougkri, Ralph T Böttcher, Joey E Lai-Cheong, Daniel B Rifkin, John A McGrath, Reinhard Fässler
ABSTRACT
Kindlin-1 is an integrin tail binding protein that controls integrin activation. Mutations in the FERMT-1 gene, which encodes for Kindlin-1, lead to Kindler syndrome in man, which is characterized by skin blistering, premature skin aging and skin cancer of unknown etiology. Here we show that loss of Kindlin-1 in mouse keratinocytes recapitulates Kindler syndrome and also produces enlarged and hyperactive stem cell compartments, which lead to hyperthickened epidermis, ectopic hair follicle development and increased skin tumor susceptibility. Mechanistically, Kindlin-1 controls keratinocyte adhesion through β1-class integrins and proliferation and differentiation of cutaneous epithelial stem cells by promoting α(v)β(6) integrin-mediated transforming growth factor-β (TGF-β) activation and inhibiting Wnt-β-catenin signaling through integrin-independent regulation of Wnt ligand expression. Our findings assign Kindlin-1 the previously unknown and essential task of controlling cutaneous epithelial stem cell homeostasis by balancing TGF-β-mediated growth-inhibitory signals and Wnt-β-catenin-mediated growth-promoting signals.
Sigma-Aldrich
IWR-1, ≥98% (HPLC)
Sigma-Aldrich
Wnt Antagonist II, IWP-2, The Wnt Antagonist II, IWP-2, also referenced under CAS 686770-61-6, controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Cancer applications.
Sigma-Aldrich
Anti-Integrin αVβ6 Antibody, clone 10D5, azide free, clone 10D5, Chemicon®, from mouse
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Kindlin-2 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution