Merck
  • Generation of stable monoclonal antibody-producing B cell receptor-positive human memory B cells by genetic programming.
Generation of stable monoclonal antibody-producing B cell receptor-positive human memory B cells by genetic programming.
Nature medicine (2009-12-22)
Mark J Kwakkenbos, Sean A Diehl, Etsuko Yasuda, Arjen Q Bakker, Caroline M M van Geelen, Michaël V Lukens, Grada M van Bleek, Myra N Widjojoatmodjo, Willy M J M Bogers, Henrik Mei, Andreas Radbruch, Ferenc A Scheeren, Hergen Spits, Tim Beaumont
ABSTRACT
The B cell lymphoma-6 (Bcl-6) and Bcl-xL proteins are expressed in germinal center B cells and enable them to endure the proliferative and mutagenic environment of the germinal center. By introducing these genes into peripheral blood memory B cells and culturing these cells with two factors produced by follicular helper T cells, CD40 ligand (CD40L) and interleukin-21 (IL-21), we convert them to highly proliferating, cell surface B cell receptor (BCR)-positive, immunoglobulin-secreting B cells with features of germinal center B cells, including expression of activation-induced cytidine deaminase (AID). We generated cloned lines of B cells specific for respiratory syncytial virus and used these cells as a source of antibodies that effectively neutralized this virus in vivo. This method provides a new tool to study B cell biology and signal transduction through antigen-specific B cell receptors and for the rapid generation of high-affinity human monoclonal antibodies.
Sigma-Aldrich
Interleukin-21 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.