Merck
Small molecule inhibition of RISC loading.
ACS chemical biology (2011-10-27)
Grace S Tan, Chun-Hao Chiu, Barry G Garchow, David Metzler, Scott L Diamond, Marianthi Kiriakidou
ABSTRACT
Argonaute proteins are the core components of the microRNP/RISC. The biogenesis and function of microRNAs and endo- and exo- siRNAs are regulated by Ago2, an Argonaute protein with RNA binding and nuclease activities. Currently, there are no in vitro assays suitable for large-scale screening of microRNP/RISC loading modulators. We describe a novel in vitro assay that is based on fluorescence polarization of TAMRA-labeled RNAs loaded to human Ago2. Using this assay, we identified potent small-molecule inhibitors of RISC loading, including aurintricarboxylic acid (IC(50) = 0.47 μM), suramin (IC(50) = 0.69 μM), and oxidopamine HCL (IC(50) = 1.61 μM). Small molecules identified by this biochemical screening assay also inhibited siRNA loading to endogenous Ago2 in cultured cells.
Sigma-Aldrich
Suramin sodium salt, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human DHX9