Merck
  • THE ROLE OF GLUCOSE 6-PHOSPHATE IN THE REGULATION OF GLUCOSE METABOLISM IN HUMAN ERYTHROCYTES.
THE ROLE OF GLUCOSE 6-PHOSPHATE IN THE REGULATION OF GLUCOSE METABOLISM IN HUMAN ERYTHROCYTES.
The Journal of biological chemistry (1964-01-01)
I A ROSE, E L O'CONNELL
Product Description
Roche
Glucose-6-phosphate, mol wt (glucose-6-P: Mr = 260.2, glucose-6-P-Na2: Mr = 304.2), pkg of 5 g