Merck
  • Secretory cargo sorting by Ca2+-dependent Cab45 oligomerization at the trans-Golgi network.
Secretory cargo sorting by Ca2+-dependent Cab45 oligomerization at the trans-Golgi network.
The Journal of cell biology (2016-05-04)
Alvaro H Crevenna, Birgit Blank, Andreas Maiser, Derya Emin, Jens Prescher, Gisela Beck, Christine Kienzle, Kira Bartnik, Bianca Habermann, Mehrshad Pakdel, Heinrich Leonhardt, Don C Lamb, Julia von Blume
ABSTRACT
Sorting and export of transmembrane cargoes and lysosomal hydrolases at the trans-Golgi network (TGN) are well understood. However, elucidation of the mechanism by which secretory cargoes are segregated for their release into the extracellular space remains a challenge. We have previously demonstrated that, in a reaction that requires Ca(2+), the soluble TGN-resident protein Cab45 is necessary for the sorting of secretory cargoes at the TGN. Here, we report that Cab45 reversibly assembles into oligomers in the presence of Ca(2+) These Cab45 oligomers specifically bind secretory proteins, such as COMP and LyzC, in a Ca(2+)-dependent manner in vitro. In intact cells, mutation of the Ca(2+)-binding sites in Cab45 impairs oligomerization, as well as COMP and LyzC sorting. Superresolution microscopy revealed that Cab45 colocalizes with secretory proteins and the TGN Ca(2+) pump (SPCA1) in specific TGN microdomains. These findings reveal that Ca(2+)-dependent changes in Cab45 mediate sorting of specific cargo molecules at the TGN.
Sigma-Aldrich
Anti-ATP2C1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)