Merck
  • Protective effects of leukemia inhibitory factor against oxidative stress during high glucose-induced apoptosis in podocytes.
Protective effects of leukemia inhibitory factor against oxidative stress during high glucose-induced apoptosis in podocytes.
Cell Stress & Chaperones (2012)
Xu J et al