Merck
  • Human farnesyl pyrophosphate synthase is allosterically inhibited by its own product.
Human farnesyl pyrophosphate synthase is allosterically inhibited by its own product.
Nature communications (2017-01-18)
Jaeok Park, Michal Zielinski, Alexandr Magder, Youla S Tsantrizos, Albert M Berghuis
ABSTRACT
Farnesyl pyrophosphate synthase (FPPS) is an enzyme of the mevalonate pathway and a well-established therapeutic target. Recent research has focused around a newly identified druggable pocket near the enzyme's active site. Pharmacological exploitation of this pocket is deemed promising; however, its natural biological function, if any, is yet unknown. Here we report that the product of FPPS, farnesyl pyrophosphate (FPP), can bind to this pocket and lock the enzyme in an inactive state. The K
Sigma-Aldrich
Isopentenyl pyrophosphate triammonium salt solution, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
Sigma-Aldrich
γ,γ-Dimethylallyl pyrophosphate triammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥90% (TLC)
Sigma-Aldrich
Geranyl pyrophosphate ammonium salt, 1 mg/mL in methanol (:aqueous 10 mM NH4OH (7:3)), ≥95% (TLC)
Sigma-Aldrich
Farnesyl pyrophosphate ammonium salt, methanol:ammonia solution, ≥95% (TLC)