Merck
  • Effect of pressure at primary drying of freeze-drying mouse sperm reproduction ability and preservation potential.
Effect of pressure at primary drying of freeze-drying mouse sperm reproduction ability and preservation potential.
Reproduction (Cambridge, England) (2007)
Kawase Y et al.