Merck
  • A G1-like state allows HIV-1 to bypass SAMHD1 restriction in macrophages.
A G1-like state allows HIV-1 to bypass SAMHD1 restriction in macrophages.
The EMBO journal (2017-01-27)
Petra Mlcochova, Katherine A Sutherland, Sarah A Watters, Cosetta Bertoli, Rob Am de Bruin, Jan Rehwinkel, Stuart J Neil, Gina M Lenzi, Baek Kim, Asim Khwaja, Matthew C Gage, Christiana Georgiou, Alexandra Chittka, Simon Yona, Mahdad Noursadeghi, Greg J Towers, Ravindra K Gupta
ABSTRACT
An unresolved question is how HIV-1 achieves efficient replication in terminally differentiated macrophages despite the restriction factor SAMHD1. We reveal inducible changes in expression of cell cycle-associated proteins including MCM2 and cyclins A, E, D1/D3 in macrophages, without evidence for DNA synthesis or mitosis. These changes are induced by activation of the Raf/MEK/ERK kinase cascade, culminating in upregulation of CDK1 with subsequent SAMHD1 T592 phosphorylation and deactivation of its antiviral activity. HIV infection is limited to these G1-like phase macrophages at the single-cell level. Depletion of SAMHD1 in macrophages decouples the association between infection and expression of cell cycle-associated proteins, with terminally differentiated macrophages becoming highly susceptible to HIV-1. We observe both embryo-derived and monocyte-derived tissue-resident macrophages in a G1-like phase at frequencies approaching 20%, suggesting how macrophages sustain HIV-1 replication in vivo Finally, we reveal a SAMHD1-dependent antiretroviral activity of histone deacetylase inhibitors acting via p53 activation. These data provide a basis for host-directed therapeutic approaches aimed at limiting HIV-1 burden in macrophages that may contribute to curative interventions.
Sigma-Aldrich
SAHA, ≥98% (HPLC)
Sigma-Aldrich
Anti-MCM2 antibody, Rabbit monoclonal, recombinant, expressed in proprietary host, clone SP85, affinity isolated antibody