Merck
  • Evolution of vocal patterns: tuning hindbrain circuits during species divergence.
Evolution of vocal patterns: tuning hindbrain circuits during species divergence.
The Journal of experimental biology (2016-12-25)
Charlotte L Barkan, Erik Zornik, Darcy B Kelley
ABSTRACT
The neural circuits underlying divergent courtship behaviors of closely related species provide a framework for insight into the evolution of motor patterns. In frogs, male advertisement calls serve as unique species identifiers and females prefer conspecific to heterospecific calls. Advertisement calls of three relatively recently (∼8.5 Mya) diverged species -
Product Description
Supelco
Serotonin, analytical standard
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid