- Determination of ethanol in beer by direct injection gas chromatography: a comparison of six identical systems
Determination of ethanol in beer by direct injection gas chromatography: a comparison of six identical systems
Journal of the Institute of Brewing (1995)
Clarkson, S.P., Ormrod, I.H.L., Sharpe, F.R.