Merck
LPS/TLR4 signal transduction pathway.
Cytokine (2008-02-29)
Yong-Chen Lu, Wen-Chen Yeh, Pamela S Ohashi
ABSTRACT
The stimulation of Toll-like receptor 4 (TLR4) by lipopolysaccharide (LPS) induces the release of critical proinflammatory cytokines that are necessary to activate potent immune responses. LPS/TLR4 signaling has been intensively studied in the past few years. Here we review molecules involved in TLR4-mediated signaling, including players that are involved in the negative regulation of this important pathway.
Sigma-Aldrich
Lipopolysaccharides from Proteus vulgaris, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharide from Porphyromonas gingivalis, purified by phenol extraction
Sigma-Aldrich
Lipopolysaccharides from Proteus mirabilis, purified by phenol extraction