Merck
  • Hypoxia-inducible microRNA-218 inhibits trophoblast invasion by targeting LASP1: Implications for preeclampsia development.
Hypoxia-inducible microRNA-218 inhibits trophoblast invasion by targeting LASP1: Implications for preeclampsia development.
The international journal of biochemistry & cell biology (2017-04-17)
Min Fang, Hechun Du, Bing Han, Guiyu Xia, Xiaoliang Shi, Feng Zhang, Qiqin Fu, Tao Zhang
ABSTRACT
Preeclampsia (PE) is a major contributor to maternal morbidity and mortality. However, the molecular mechanisms underlying PE progression are not well characterized. Here, we investigated the role of miR-218 in PE development. The expression of miR-218 and its host genes SLIT2 and SLIT3 was up-regulated in preeclamptic placentae compared to normal placentae. miR-218 expression was induced by hypoxia and decreased after knockdown of HIF-1α in an extravillous trophoblast cell line (HTR-8/SVneo). Chromatin immunoprecipitation assays showed direct binding of HIF-1α to the promoters of SLIT2 and SLIT3. Bioinformatics analysis identified LASP1 as a direct target of miR-218. Overexpression of miR-218 repressed the expression of LASP1 at both the mRNA and protein level. Meanwhile, miR-218 repressed the activity of a luciferase reporter containing the 3'-untranslated region of the LASP1 gene. Furthermore, expression of LASP1 rescued the inhibitory effect of miR-218 on HTR-8/SVneo cell invasion. Together, these results indicated that miR-218 contributes to PE by targeting LASP1 to inhibit trophoblast invasion.
Sigma-Aldrich
MISSION® esiRNA, targeting human LASP1