Merck
  • A20/TNFAIP3 Discriminates Tumor Necrosis Factor (TNF)-Induced NF-κB from JNK Pathway Activation in Hepatocytes.
A20/TNFAIP3 Discriminates Tumor Necrosis Factor (TNF)-Induced NF-κB from JNK Pathway Activation in Hepatocytes.
Frontiers in physiology (2017-09-08)
Federico Pinna, Michaela Bissinger, Katharina Beuke, Nicolas Huber, Thomas Longerich, Ursula Kummer, Peter Schirmacher, Sven Sahle, Kai Breuhahn
ABSTRACT
In the liver tumor necrosis factor (TNF)-induced signaling critically regulates the immune response of non-parenchymal cells as well as proliferation and apoptosis of hepatocytes
Sigma-Aldrich
MISSION® esiRNA, targeting human TNFAIP3