Merck
  • Androgen Receptor Pathway-Independent Prostate Cancer Is Sustained through FGF Signaling.
Androgen Receptor Pathway-Independent Prostate Cancer Is Sustained through FGF Signaling.
Cancer cell (2017-10-11)
Eric G Bluemn, Ilsa M Coleman, Jared M Lucas, Roger T Coleman, Susana Hernandez-Lopez, Robin Tharakan, Daniella Bianchi-Frias, Ruth F Dumpit, Arja Kaipainen, Alexandra N Corella, Yu Chi Yang, Michael D Nyquist, Elahe Mostaghel, Andrew C Hsieh, Xiaotun Zhang, Eva Corey, Lisha G Brown, Holly M Nguyen, Kenneth Pienta, Michael Ittmann, Michael Schweizer, Lawrence D True, David Wise, Paul S Rennie, Robert L Vessella, Colm Morrissey, Peter S Nelson
ABSTRACT
Androgen receptor (AR) signaling is a distinctive feature of prostate carcinoma (PC) and represents the major therapeutic target for treating metastatic prostate cancer (mPC). Though highly effective, AR antagonism can produce tumors that bypass a functional requirement for AR, often through neuroendocrine (NE) transdifferentiation. Through the molecular assessment of mPCs over two decades, we find a phenotypic shift has occurred in mPC with the emergence of an AR-null NE-null phenotype. These "double-negative" PCs are notable for elevated FGF and MAPK pathway activity, which can bypass AR dependence. Pharmacological inhibitors of MAPK or FGFR repressed the growth of double-negative PCs in vitro and in vivo. Our results indicate that FGF/MAPK blockade may be particularly efficacious against mPCs with an AR-null phenotype.
Sigma-Aldrich
Anti-MAP Kinase, Activated (Diphosphorylated ERK-1&2) antibody, Mouse monoclonal, clone MAPK-YT, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, ERK-2) antibody produced in rabbit, whole antiserum
Sigma-Aldrich
R1881, ≥98% (HPLC)
Sigma-Aldrich
Anti-MAP Kinase Kinase (MEK, MAPKK) antibody produced in rabbit, whole antiserum