Merck
  • Arginase Activity in Human Breast Cancer Cell Lines: N?-Hydroxy-l-arginine Selectively Inhibits Cell Proliferation and Induces Apoptosis in MDA-MB-468 Cells
Arginase Activity in Human Breast Cancer Cell Lines: N?-Hydroxy-l-arginine Selectively Inhibits Cell Proliferation and Induces Apoptosis in MDA-MB-468 Cells
Cancer Research (2000)
Rajan Singh