Merck
  • Palmitate Conditions Macrophages for Enhanced Responses toward Inflammatory Stimuli via JNK Activation.
Palmitate Conditions Macrophages for Enhanced Responses toward Inflammatory Stimuli via JNK Activation.
Journal of immunology (Baltimore, Md. : 1950) (2017-10-25)
Marta Riera-Borrull, Víctor D Cuevas, Bárbara Alonso, Miguel A Vega, Jorge Joven, Elena Izquierdo, Ángel L Corbí
ABSTRACT
Obesity is associated with low-grade inflammation and elevated levels of circulating saturated fatty acids, which trigger inflammatory responses by engaging pattern recognition receptors in macrophages. Because tissue homeostasis is maintained through an adequate balance of pro- and anti-inflammatory macrophages, we assessed the transcriptional and functional profile of M-CSF-dependent monocyte-derived human macrophages exposed to concentrations of saturated fatty acids found in obese individuals. We report that palmitate (C16:0, 200 μM) significantly modulates the macrophage gene signature, lowers the expression of transcription factors that positively regulate IL-10 expression (MAFB, AhR), and promotes a proinflammatory state whose acquisition requires JNK activation. Unlike LPS, palmitate exposure does not activate STAT1, and its transcriptional effects can be distinguished from those triggered by LPS, as both agents oppositely regulate the expression of CCL19 and
Sigma-Aldrich
Bovine Serum Albumin, fatty acid free, low endotoxin, lyophilized powder, BioReagent, suitable for cell culture, ≥96% (agarose gel electrophoresis)
Sigma-Aldrich
Sodium palmitate, ≥98.5%
Sigma-Aldrich
Arachidonic acid sodium salt from Mortierella alpina, ≥98.5% (GC)