Merck
  • SET/I2PP2A overexpression induces phenotypic, molecular, and metabolic alterations in an oral keratinocyte cell line.
SET/I2PP2A overexpression induces phenotypic, molecular, and metabolic alterations in an oral keratinocyte cell line.
The FEBS journal (2017-06-22)
Lays M Sobral, Ricardo D Coletta, Luciane C Alberici, Carlos Curti, Andréia M Leopoldino
ABSTRACT
The multifunctional SET/I2PP2A protein is known to be overexpressed in head and neck squamous cell carcinoma. However, SET has been reported to have apparently conflicting roles in promoting cancer cell survival under oxidative stress conditions and preventing invasion and metastasis, complicating efforts to understand the contribution of SET to carcinogenesis. In the present study, we overexpressed SETin a spontaneously immortalized oral keratinocyte cell line (NOK-SI SET) and demonstrated that SET upregulation alone was sufficient to transform cells. In comparison with NOK-SI cells, NOK-SI SET cells demonstrated increased levels of phosphorylated Akt, c-Myc and inactive/phosphorylated Rb, together with decreased total Rb protein levels. In addition, NOK-SI SET cells presented the following: (a) a spindle-cell shape morphology compared with the polygonal morphology of NOK-SI cells; (b) loss of mesenchymal stem cell markers CD44 and CD73, and epithelial cell markers CD71 and integrin α6/β4; (c) the ability to form xenograft tumors in nude mice; and (d) increased mitochondrial respiration accompanied by decreased ROSlevels. Overall, our results show that SEToverexpression promotes morphological and oncogenic cell transformation of an oral keratinocyte cell.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (HPLC), powder
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
Anti-MAP Kinase (ERK-1, ERK-2) antibody produced in rabbit, whole antiserum