- 5-HT modulates the rat mesenteric vasopressor outflow by 5-HT
5-HT modulates the rat mesenteric vasopressor outflow by 5-HT
5-hydroxytryptamine (5-HT) modulates noradrenergic activity in different cardiovascular territories, but its effect on the mesenteric vasopressor outflow has not yet been clarified. This study investigated the in vivo serotonergic influence, characterizing 5-HT receptors implicated, in sympathetic innervation of mesenteric vasculature. Wistar rats were anaesthetised and prepared for the in situ autoperfused rat mesentery, monitoring systemic blood pressure (SBP), heart rate (HR) and mesenteric perfusion pressure (MPP). Electrical stimulation of mesenteric sympathetic nerves resulted in frequency-dependent increases in MPP (9 ± 1.6, 25.7 ± 3.9 and 60.2 ± 5 mmHg for 2, 4 and 8 Hz, respectively), without altering SBP or HR. 5-HT (1-25 μg/kg), 5-carboxamidotryptamine (5-HT