- RNF146 Inhibits Excessive Autophagy by Modulating the Wnt-β-Catenin Pathway in Glutamate Excitotoxicity Injury.
RNF146 Inhibits Excessive Autophagy by Modulating the Wnt-β-Catenin Pathway in Glutamate Excitotoxicity Injury.
Frontiers in cellular neuroscience (2017-03-23)
Yuefan Yang, Peng Luo, Haoxiang Xu, Shuhui Dai, Wei Rao, Cheng Peng, Wenke Ma, Jiu Wang, Hongyu Xu, Lei Zhang, Sai Zhang, Zhou Fei
PMID28321181
ABSTRACT
Glutamate induced excitotoxicity is common in diverse neurological disorders. RNF146 as an E3 ubiquitin ligase protects neurons against excitotoxicity
Sigma-Aldrich
ANTI-BECLIN1-BH3 DOMAIN antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-LC3 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-p62/SQSTM1 antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution