- L-cysteine supplementation attenuates local inflammation and restores gut homeostasis in a porcine model of colitis
L-cysteine supplementation attenuates local inflammation and restores gut homeostasis in a porcine model of colitis
Biochim. Biophys. Acta Gen. Subj. (2009)
Kim CJ