Merck
  • Chemical and Sensory Evaluation of Silicone and Polylactic Acid-Based Remedial Treatments for Elevated Methoxypyrazine Levels in Wine.
Chemical and Sensory Evaluation of Silicone and Polylactic Acid-Based Remedial Treatments for Elevated Methoxypyrazine Levels in Wine.
Molecules (Basel, Switzerland) (2016-09-21)
Andreea Botezatu, Belinda S Kemp, Gary J Pickering
ABSTRACT
Alkylmethoxypyrazines (MPs) are a class of compounds that can elicit undesirable aroma and flavor characteristics in wine, and resist remediation using traditional wine making approaches. MPs are grape-derived constituents as well as contaminants from Coccinellidae beetles present during wine processing; the latter eliciting an off-flavor referred to as 'ladybug taint'. In this study we investigated the capacity of two plastic polymers-one silicone-based, the other polylactic acid-based-applied with varying surface areas to reduce concentrations of isopropylmethoxypyrazine (IPMP), sec-butylmethoxypyrazine (SBMP) and isobutylmethoxypyrazine (IBMP) in a Merlot wine using multi-dimensional gas chromatography coupled with mass spectrometry and headspace solid phase microextraction (SPME-MDGCMS). The impact of treatments on the sensory characteristics of the wine (descriptive analysis) and volatile aroma compounds (VOCs) (SPME-MDGCMS) was also investigated. Results showed substantial reductions for all of the target odorants: up to 38%, 44% and 39% for IPMP, SBMP and IBMP, respectively, for the silicone polymer, and up to 75%, 78% and 77% for IPMP, SBMP and IBMP, respectively, for the polylactic acid polymer. These polymers had no or minimal effect on VOCs at applications of 200 cm²/L for silicone or for all polylactic acid treatments. Sensory impacts were less clear, but generally showed minimal effect from the treatments. Taken overall, the data confirm the utility of both polylactic acid and silicone polymers in reducing elevated levels of grape-derived MPs, as well as potentially improving wine contaminated by ladybug taint.
Product Description
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #2, Fused Silica (1 cm), size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3
Supelco
SPME Fiber Assembly Kit, Assortment #5, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPME Fiber Assembly, 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Sampling Stand, for use with 15 mL vials
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 23 ga, Autosampler, pk of 3, notched gray hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, light blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME Fiber Assembly, 85 μm CAR/PDMS, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, light blue hub
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, Metal Alloy (1cm), needle size 23 ga, Autosampler, pk of, plain gray hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 1, pink hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, green hub
Supelco
SPME Fiber Assembly, 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPME Fiber Assembly, 7 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, green hub