Merck
  • Benzo[b]thiophene-2-carboxamide derivatives as potent urotensin-II receptor antagonists.
Benzo[b]thiophene-2-carboxamide derivatives as potent urotensin-II receptor antagonists.
Bioorganic & medicinal chemistry letters (2016-09-07)
Chae Jo Lim, Seong Eun Woo, Su Ik Ko, Byung Ho Lee, Kwang-Seok Oh, Kyu Yang Yi
ABSTRACT
Members of a series of benzo[b]thiophene-2-carboxamide derivatives, possessing an N-(1-(3-bromo-4-(piperidin-4-yloxy)benzyl)piperidin-4-yl) group, were synthesized and evaluated as urotensin-II receptor antagonists. The results show that these substances have potent UT binding affinities. Observations made in a systematic SAR investigation of the effects of a variety of substituents (R(1) and R(2)) at the 5- and 6-positions in the benzo[b]thiophene-2-carboxamide moiety on UT binding affinities led to identification of the 5-cyano analog 7f as a highly potent UT antagonist with an IC50 value of 25nM. Despite having a good metabolic stability, 7f is a potent inhibitor of CYP isozyme and displays an unsuitable PK profile.
Product Description
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® Cyano Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm