Merck
LC/MS Analysis of Fructose and NaCl on SeQuant® ZIC-pHILIC
LC/MS Analysis of Fructose and NaCl on SeQuant® ZIC-pHILIC application for HPLC
CONDITIONS
column
SeQuant® ZIC-pHILIC, PEEK, 150 x 4.6 mm, 5 μm, 100 Å (1.50461.0001)
mobile phase
[A] acetonitrile; [B] ammonium acetate 100 mM pH 5; (80:20, A:B)
flow rate
1.5 mL/min
pressure
2000 psi (135 bar)
column temp.
40 °C
detector
Sedere Sedex 85 ELSD (cell: analytical nebulizer)
injection
20 μL
sample
1000 ppm Fructose & 90ppm NaCl
Description
Legal Information
SeQuant is a registered trademark of Merck KGaA, Darmstadt, Germany