Merck
HomeDetermination of Neonicotinoids in Honey using a Chromolith RP-18 HPLC column and UV Detection
Determination of Neonicotinoids in Honey using a Chromolith RP-18 HPLC column and UV Detection
Anita Piper
Life Science Application Centre (GAC), Darmstadt, Germany
Introduction
Neonicotinoids are water soluble insecticides. They are used for crop protection and can migrate from treated soil into plant tissue and flower nectar. Neonicotinoids have a negative effect on brain cells of honey bees, and colony collapse can occur. [Clothianidin, Imidacloprid and Thiamethoxam are banned in Germany, (EU) No. 485/2013].
This application focuses on HPLC testing of neonicotinoids in honey. The limit of detection (LOD) and limit of quantitation (LOQ) for neonicotinoids is better than 30 µM and 90 µM, respectively, when using HPLC combined with UV detection.
Determination of Neonicotinoids in Honey
Chromatographic Data - blank run
Chromatographic Data - blank run
Chromatographic Data - blank run
Chromatographic Data - Standard Solution
Standard Solution (200 µM @ 254 nm)
Standard Solution (200 µM @ 254 nm)
Standard Solution (200 µM @ 270 nm)
Standard Solution (200 µM @ 270 nm)
Chromatographic Data - Sample Solution spiked
Sample Solution Spiked @ 254 nm
Sample Solution Spiked @ 254 nm
Sample Solution @ 270 nm
 Sample Solution @ 270 nm
Chromatographic Data - Sample Solution spiked
Sample Solution Spiked @ 254 nm
Sample Solution @ 254 nm
Sample Solution Spiked @ 270 nm
Sample Solution Spiked @ 270 nm
1. Specificity: Inject standard solution of neonicotinoids (200 µM) and determine the retention time and content of desired analyte @254 nm
1. Specificity: Inject standard solution of neonicotinoids (200 µM) and determine the retention time and content of desired analyte @270 nm
2. Standard repeatability honey (spiked with 5 µL of a 200 µM standard solution) in mAU*min
3. LOD & LOQ
4. Linearity (area mAU*min)
Dinutefuran @270 nm
Dinutefuran
Nitenpyram @270 nm
Nitenpyram
Thiamethoxam @254 nm
Thiamethoxam
Imidacloprid @270 nm
Imidacloprid
Clothianidin @270 nm
Clothianidin
Acetamiprid @254 nm
Acetamiprid
Thiacloprid @254 nm
Thiacloprid
Materials List