Merck
HomeViable Air MonitoringMicrobial Air Samplers for Isolators in the Pharmaceutical Industry
Microbial Air Samplers for Isolators in the Pharmaceutical Industry
The highest standards and quality in a pharmaceutical production environment are directly linked to the microbiological safety of the finished products. Ensure active microbial air sampling solutions generate precise, reliable, and reproducible results with our easy-to-use environmental monitoring systems that fully comply with international standards and regulations.
MAS-100® MICROBIAL AIR SAMPLERS FOR ENVIRONMENTAL MONITORING
The MAS-100® air samplers are designed for simple and reliable microbial monitoring of ambient air and compressed gas in pharmaceutical manufacturing facilities. The working of all the MAS-100® air monitoring systems is based on the Anderson impaction principle, where the air is aspirated through a perforated lid.
Mas®-100 microbial air samplers for environmental monitoring
  • Flow rate of 100 L/min
  • Precise, reproducible measurements and outstanding user flexibility
  • Excellent collection efficiency
  • Fully automatic calibration
  • Validation documents in accordance with ISO 14698 part 1 and part 2
MICROBIAL AIR SAMPLERS FOR ISOLATORS
The MAS-100® Iso series of integrated microbial air monitoring systems serve to sample and monitor isolators and restricted access barrier systems (RABS) for airborne organisms with the greatest accuracy. The system is applicable when production processes are subject to the most stringent requirements. Discover our most accurate microbial air samplers for use in isolators:
MAS-100 ISO MH® AIR SAMPLER FOR ISOLATORS AND RABS
MAS-100 Iso MH® air sampler for isolators and RABS
The MAS-100 Iso MH® air sampler is used to control the microbiological contamination of the air in isolators. A unique security concept permits the installation of the sampling heads for standard 90–100 mm Petri dishes at the critical control points. All electronic and moving parts are outside the critical zone. The MAS-100 Iso MH® has an additional internal pump with flow control for automatic disinfection of the sampling head and the aspiration tube.
  • Operates up to 4 sampling heads with one pump
  • Innovative double-valve system enables the sampling heads to be integrated into the decontamination process of the isolator
  • Each unit has both an air and a sterilization pump which work fully autonomously
  • Built according to GAMP 5 and corresponds to the ISO 14698 standard
  • Independently validated with ICR settle plates for physical and biological efficiency
  • Designed for the highest demands
The MAS 100 Iso MH® can support up to 30 meters of tube from the control unit to each head and the control unit is IP 64 as standard.
MAS-100 Iso MH® System and Accessories
Loading
MAS-100 ISO NT® MICROBIAL AIR SAMPLER FOR ASEPTIC PRODUCTION AND STERILITY TESTING ISOLATORS
MAS-100 Iso NT® air sampler for use in aseptic production and sterility testing isolators
The MAS-100 Iso NT® air sampler is specifically designed for use in aseptic production and sterility testing isolators. The system operates with an integrated mass flow sensor to ensure a constant flow rate of 100 L/min and uses 90–100 mm Petri dishes . The sampling head is mounted inside the isolator, and all electronic and moving parts remain outside the critical zone.
  • Produced to GAMP 4 specifications and is compliant with ISO 14698 guidelines
  • Double valve system prevents contaminants from entering the isolator
  • Automatic in-line decontamination of the sampling head and aspiration tube
  • Ethernet, Profibus and 9 digital inputs/outputs
  • IP 54 and IP 66 models are available
  • Allows for remote activation via a computer or the PLC controlled on the isolator.
MAS-100 Iso NT® System and Accessories
Loading
Maintenance, Validation & Qualification Services
We offer comprehensive services to help you save your QC resources and facilitate the implementation of your MAS-100 Iso MH® and MAS-100 Iso NT® air samplers into your daily testing routine. Our highly experienced teams are there to assist you.